Learn More
Selenium(IV) oxide, 99.8%, (trace metal basis)
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 315.0°C |
| Color | White to Beige |
| Physical Form | Powder |
| Quantity | 2.5 kg |
| Assay Percent Range | 70 to 71.5% Se |
| Linear Formula | SeO2 |
| Fieser | 01,992; 02,360; 03,245; 04,422; 05,575; 06,509; 08,439; 09,409; 10,345; 11,96; 13,272; 14,177; 15,89; 17,312 |
| Merck Index | 15,8571 |
| Solubility Information | Solubility in water: 38.4 g/100mL (14°C). Other solubilities: 1.11 parts/100 parts acetic acid (13.9°C), 10.16 parts/100 parts methanol (11.8°C), 6.67 parts/100 parts 93% ethanol (14°C), 4.35 parts/100 parts acetone (15.3°C) |
| IUPAC Name | selenium dioxide |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| EINECSNumber | 231-194-7 |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.