Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Selenium, 99.5+%, powder, -200 mesh

Catalog No. p-4509808
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle

CAS: 7782-49-2 | Se | 78.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-49-2
Molecular Formula Se
Molecular Weight (g/mol) 78.97
MDL Number MFCD00134090 MFCD00011224
InChI Key BUGBHKTXTAQXES-UHFFFAOYSA-N
Synonym selenium, elemental, selen, hydride, atom, elemental, selenio, vandex, alloy, and compounds, gray
PubChem CID 6326970
ChEBI CHEBI:27568
IUPAC Name selenium
SMILES [Se]

Specifications

Boiling Point 685.0°C
Melting Point 217.0°C
Color Colorless to Yellow
Physical Form Solution
Quantity 50 g
Assay Percent Range 99.5% min
Linear Formula Se
Fieser 01,990; 04,422; 05,575; 06,507; 08,439; 10,345; 11,465; 14,281
Merck Index 158,568
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in concentrated acid, slightly soluble in diehtyl ether, almost insoluble in carbon disulfide, insoluble in ethanol
Formula Weight 78.96
Percent Purity ≥99.5%
Packaging Glass bottle
Specific Gravity 4.81
Density 4.8100 g/mL
Chemical Name or Material Selenium
Show More Show Less
Hazard Category GHS Signal Word: Danger
EINECSNumber 231-957-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.