Selenium Powder, -325 mesh, 99.5% (Metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 1060106

Quantity 5g

 View more versions of this product

Catalog No. AA1060106

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7782-49-2
    Physical Form Powder
    Assay Percent Range 99.5% (Metals basis)
    MDL Number MFCD00134090
    Chemical Name or Material Selenium

    Specifications

    Quantity 5g
    Color Various
    DOT Information DOT Class: 9, Packaging Group: III
    Density 4.28g/cm3
    EINECS Number 231-957-4
    Physical Form Powder
    Odor Odorless
    Assay Percent Range 99.5% (Metals basis)
    Appearance Various colors powder
    MDL Number MFCD00134090
    CAS 7782-49-2
    Health Hazard 1 H301-H331-H373-H413: Toxic if swallowed. Toxic if inhaled. May cause damage to organs through prolonged or repeated exposure. May cause long lasting harmful effects to aquatic life.
    Health Hazard 2 When using do not eat, drink or smoke. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). Avoid release to the environment. Refer to special instructions/Safety data sheets
    Chemical Name or Material Selenium

    When using do not eat, drink or smoke. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). Avoid release to the environment. Refer to special instructions/Safety data sheets

    H301-H331-H373-H413: Toxic if swallowed. Toxic if inhaled. May cause damage to organs through prolonged or repeated exposure. May cause long lasting harmful effects to aquatic life.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit