Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Selenium standard solution, for AAS, 1 mg/ml Se in 10% HCl

Catalog No. p-220872
Click to view available options
Quantity:
100 mL
500 mL

CAS: 7782-49-2 | Se | 78.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for atomic adsorption spectroscopy
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Selenium standard solution
Name Note 1 mg/ml Se in 10% HCl
Packaging Plastic bottle
CAS 7782-49-2, 7647-01-0, 7732-18-5
Density 4.81
Physical Form Solution
Molecular Formula Se
Concentration 995 to 1005 μg/mL of Se in 10% HCl (20°C)
MDL Number MFCD00134090 MFCD00011224
Merck Index 14, 8431
Solubility Soluble in water
InChI Key BUGBHKTXTAQXES-UHFFFAOYSA-N
SMILES [Se]
IUPAC Name selenium
Molecular Weight (g/mol) 78.97
Quantity 100 mL
PubChem CID 6326970
ChEBI CHEBI:27568
Formula Weight 78.96
Grade AAS
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.