Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Selenium sulfide, 94%

Catalog No. AA4294336
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Catalog No. Quantity
AA4294336 500 g
AA4294314 25 g
AA4294322 100 g
3 options

Catalog No. AA4294336

Supplier: Thermo Scientific Chemicals 04294336

Only null left
Add to Cart

CAS: 7488-56-4 | S2Se | 143.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7488-56-4
Molecular Formula S2Se
Molecular Weight (g/mol) 143.09
MDL Number MFCD00011227
InChI Key JNMWHTHYDQTDQZ-UHFFFAOYSA-N
Synonym selenium disulfide, selenium disulphide, selsun blue, selenium iv disulfide, selenium sulfide ses2, sebusan, selenex, selenix, selukos, lenium
PubChem CID 24087
IUPAC Name thione
SMILES S=[Se]=S

Specifications

Boiling Point (decomposition)
Melting Point <100°C
Color Red to Brown
Physical Form Powder
Quantity 500 g
Assay Percent Range 94%
Solubility Information Insoluble in water,in dilute HCl
Formula Weight 143.09
Chemical Name or Material Selenium sulfide
Hazard Category H301+H331-H373
Precautionary Statement P262-P264b-P270-P280-P301+P310-P304+P340-P311-P314-P330-P501c
DOTInformation DOT Class: 6.1; Packaging Group: III
EINECSNumber 231-303-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.