Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Seleno-L-methionine

Catalog No. p-7057565
Click to view available options
Quantity:
100 mg
500 mg
1 g

CAS: 3211-76-5 | C5H11NO2Se | 196.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3211-76-5
Molecular Formula C5H11NO2Se
Molecular Weight (g/mol) 196.12
MDL Number MFCD00037210
InChI Key RJFAYQIBOAGBLC-BYPYZUCNSA-N
Synonym l-selenomethionine, l-+-selenomethionine, selenium-l-methionine, l-selenomethioninum, seleno-l-methionine, s-2-amino-4-methylselanyl butanoic acid, semet, s-2-amino-4-methylseleno butyric acid, unii-964mrk2pel, s-2-amino-4-methylseleno butanoic acid
PubChem CID 105024
ChEBI CHEBI:62621
IUPAC Name (2S)-2-amino-4-methylselanylbutanoic acid
SMILES C[Se]CC[C@H](N)C(O)=O

Specifications

Melting Point 265°C
Color White
Linear Formula CH3SeCH2CH2CH(NH2)CO2H
UN Number UN3283
Solubility Information Soluble in water
Quantity 100 mg
Formula Weight 196.11
Physical Form Powder
Chemical Name or Material Seleno-L-methionine
Hazard Category H301+H331-H373
Hazard Statement GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
RTECSNumber EK7713840
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.