Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Seleno-L-methionine

Click to view available options
Quantity:
100mg
500mg
1g
3211-76-5
C5H11NO2Se
196.119
MFCD00037210
RJFAYQIBOAGBLC-BYPYZUCNSA-N
l-selenomethionine,l-+-selenomethionine,selenium-l-methionine,l-selenomethioninum,seleno-l-methionine,s-2-amino-4-methylselanyl butanoic acid,semet,s-2-amino-4-methylseleno butyric acid,unii-964mrk2pel,s-2-amino-4-methylseleno butanoic acid
105024
CHEBI:62621
(2S)-2-amino-4-methylselanylbutanoic acid
C[Se]CCC(C(=O)O)N
3211-76-5
C5H11NO2Se
196.119
MFCD00037210
RJFAYQIBOAGBLC-BYPYZUCNSA-N
l-selenomethionine,l-+-selenomethionine,selenium-l-methionine,l-selenomethioninum,seleno-l-methionine,s-2-amino-4-methylselanyl butanoic acid,semet,s-2-amino-4-methylseleno butyric acid,unii-964mrk2pel,s-2-amino-4-methylseleno butanoic acid
105024
CHEBI:62621
(2S)-2-amino-4-methylselanylbutanoic acid
C[Se]CCC(C(=O)O)N
Linear Formula CH3SeCH2CH2CH(NH2)CO2H
UN Number UN3283
Formula Weight 196.11
Physical Form Powder
Color White
Melting Point 265°C
Quantity 100mg
Chemical Name or Material Seleno-L-methionine

GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: II; Proper Shipping Name: SELENIUM COMPOUND, SOLID, N.O.S.

RTECSNumber : EK7713840

storageNote1 : -30° to -10°C