Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Selenoprotein W Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567614
Description
Selenoprotein W Polyclonal Antibody for IHC (P)
This gene encodes a selenoprotein containing a selenocysteine (Sec) residue, which is encoded by the UGA codon that normally signals translation termination. The 3' UTRs of selenoprotein mRNAs contain a conserved stem-loop structure, the Sec insertion sequence (SECIS) element that is necessary for the recognition of UGA as a Sec codon rather than as a stop signal. This protein is highly expressed in skeletal muscle, heart and brain. It belongs to the SelWTH family, which possesses a thioredoxin-like fold and a conserved CxxU (C is cysteine, U is Sec) motif, suggesting a redox function for this gene. Studies in mouse show that this selenoprotein is involved in muscle growth and differentiation, and in the protection of neurons from oxidative stress during neuronal development. A retroprocessed pseudogene for this locus has been identified on chromosome 1.
Specifications
Selenoprotein W | |
Polyclonal | |
Unconjugated | |
SELENOW | |
Selenoprotein W; selenoprotein W, 1; selenoprotein W, muscle 1; Selenow; SelW; Sepw1 | |
Rabbit | |
affinity chromatography | |
RUO | |
6415 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.8 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P63302 | |
SELENOW | |
Synthesized peptide derived from human Selenoprotein W. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction