Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Selenourea, 98+%

Catalog No. p-3737340
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

CAS: 630-10-4 | CH4N2Se | 123.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 630-10-4
Molecular Formula CH4N2Se
Molecular Weight (g/mol) 123.02
MDL Number MFCD00008065
InChI Key ZUCFIOXGLBMWHV-UHFFFAOYSA-N
Synonym selenourea, 2-selenourea, carbamimidoselenoic acid, selenouronium, urea, seleno, rcra waste number p103, rcra waste no. p103, iykvlicpfcezof-uhfffaoysa-n, ch4n2se, isoselenourea
PubChem CID 6327594
IUPAC Name $l^{1}-selanylmethanimidamide
SMILES C(=N)(N)[Se]

Specifications

Melting Point 214.0°C to 215.0°C
Color Gray to Pink
Infrared Spectrum Authentic
Assay Percent Range 98% min
Packaging Glass bottle
Linear Formula H2NC(=Se)NH2
Quantity 1 g
Solubility Information Solubility in water: very soluble.
Formula Weight 123.02
Percent Purity 98+%
Physical Form Crystalline Powder or Needles
Chemical Name or Material Selenourea
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
EINECSNumber 211-129-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.