Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SELH Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561721
Description
Immunogen sequence: KVNPTKPRRG SFEVTLLRPD GSSAELWTGI KKGPPRKLKF PEPQEVVEEL KKYLS Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 87%.

Specifications
SELH | |
Polyclonal | |
Unconjugated | |
SELENOH | |
C11orf31; C17orf10; chromosome 11 open reading frame 31; SELENOH; Selenoprotein H; SELH | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
280636 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.10 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IZQ5 | |
SELENOH | |
Recombinant protein corresponding to Human SELH. Recombinant protein control fragment (Product #RP-100474). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction