Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SELK Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP326091PE
Description
SELK Polyclonal antibody specifically detects SELK in Human samples. It is validated for Western Blot, ImmunocytochemistrySpecifications
SELK | |
Polyclonal | |
Western Blot, Immunocytochemistry | |
HSPC297, MGC17057, selenoprotein K, SelK | |
Produced in rabbits immunized with E. coli-derived Human SELK fragment | |
0.1 mL | |
Innate Immunity | |
58515 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunocytochemistry | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction