Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SELK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578549
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578549 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578549 Supplier Invitrogen™ Supplier No. PA578549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: K562 Predicted Reactivity: Dog (97%), Pig (91%), Rabbit (89%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (89%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

SELK is a selenoprotein, containing a selenocysteine residue at its active site. SELK is localized to the endoplasmic reticulum and is highly expressed in the heart, where it may function as an antioxidant. SELK can bind to proteins such as the ER-associated degradation (ERAD) components p97 ATPase, Derlin-1 and -2, and SelS as well as proteins in the oligosaccharyltransferase complex, suggesting that SELK may be involved in the Derlin-dependent ERAD of glycosylated misfolded proteins. SELK is also thought to be important in Ca2+ flux in immune cells, and is cleaved by m-calpain in resting macrophages. When these macrophages are activated through different Toll-like receptors, this cleavage is inhibited.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SELK
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.48 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene SELENOK
Gene Accession No. Q9Y6D0
Gene Alias 1110001C03Rik; AA673253; heat shock protein; heat shock protein, 30 kDa; Hsp30; HSPC030; HSPC297; Selenok; Selenoprotein K; SELK
Gene Symbols SELENOK
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human SELK. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58515
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.