Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SELK Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$148.20 - $378.00
Specifications
Antigen | SELK |
---|---|
Dilution | Western Blot 1:500 - 1:2000 |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
SELK Polyclonal antibody specifically detects SELK in Human, Mouse samples. It is validated for Western BlotSpecifications
SELK | |
Western Blot | |
Unconjugated | |
Rabbit | |
Innate Immunity | |
PBS, 50% glycerol, pH7.3 | |
58515 | |
IgG | |
Affinity purified |
Western Blot 1:500 - 1:2000 | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse | |
HSPC297, MGC17057, selenoprotein K, SelK | |
A synthetic peptide corresponding to a sequence within amino acids 1-94 of human SELK (NP_067060.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title