Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SELL Mouse anti-Human, Clone:8C8B7, Abnova™

Catalog No. 89333181 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-181 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-181 Supplier Abnova Corporation Supplier No. MAB16689
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human SELL.

SELL is a cell surface component that is a member of a family of adhesion/homing receptors which play important roles in leukocyte-endothelial cell interactions. The molecule is composed of multiple domains: one homologous to lectins, one to epidermal growth factor, and two to the consensus repeat units found in C3/C4 binding proteins. [provided by RefSeq]

Specifications

Antigen SELL
Applications ELISA, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 8C8B7
Conjugate Unconjugated
Description selectin L
Dilution ELISA (1:10000)
Western Blot (1:500-1:2000)
Immunohistochemistry
Immunocytochemistry (1:200-1:1000)
Flow Cytometry
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene SELL
Gene Alias CD62L, LAM-1, LAM1, LECAM1, LNHR, LSEL, LYAM1, Leu-8, Lyam-1, PLNHR, TQ1, hLHRc
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acid 83-186 of human SELL from E. coli.
Quantity 100 μg
Gene ID (Entrez) 6402
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.