Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Sell (Rat) ELISA Kit
Click to view available options
Detection Limit:
<20 pg/ml
6 pg/mL
Specifications
Specifications
Calibration Range | 625 to 40000 pg/mL |
Content And Storage | Store the kit at -20°C |
Description | Sell (Rat) is a sandwich enzyme immunoassay for the quantitative measurement of rat Sell. |
Conjugate | Biotin |
Cross Reactivity | Rat |
Detection Method | Colorimetric |
Product Type | SELL (Rat) ELISA kit |
Sample Type | Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum |
Sufficient For | 96 |
For Use With (Application) | Quantification |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction