Learn More
Sellstrom™ Impact Goggles
Built for comfortable protection
Supplier:  Sellstrom™ 88000
 View more versions of this product
Catalog No. 17-288A
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
  • Clear UV-absorbing polycarbonate lens
  • Soft vinyl goggle frame conforms to facial contours for a secure fit
  • Protect from flying particles
  • Fit comfortably over most prescription eyewear
  • Direct venting ensures airflow and cools the eye area
Note:Do not use these goggles for splash protection.
Order Info
Available in standard or fog-free version.
Specifications
Specifications
Uncoated
Impact Goggles
Vinyl
Clear
Polycarbonate
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.