Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SELM Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA581005
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human SELM/Selenoprotein M.
A growing body of evidence relates selenium to cancer prevention, immune system function, male fertility, cardiovascular disorder, control of the aging and neurodiseases process. Selenoproteins are thought to be responsible for the majority of these biomedical effects of selenium. Approximately 17 selenoproteins have been identified until now. Although the function of many selenoproteins are unknown, some play important roles in anti-oxidant mechanisms. It has been also implicated in the regulation of signaling pathways through catalysis of thiol/disulfide exchange. Selenoprotein M (SelM) is especially expressed in a mammalian brain and is localized to the perinuclear structures (Golgi/ER). The roles of selM have not been clearly identified until present time.
Specifications
SELM | |
Polyclonal | |
Unconjugated | |
SELENOM | |
cb693; fc13f12; SELENOM; Selenoprotein M; selenoprotein M precursor; selenoprotein SelM; selm; SelM protein; sepm; sepm protein; wu:fc13f12; zgc:103431 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
140606 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with no preservative; pH 7 | |
Q8WWX9 | |
SELENOM | |
Recombinant Human SELM/Selenoprotein M protein (Ala24-Leu145). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction