Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Selpercatinib
GREENER_CHOICE

Catalog No. 769610
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-961-0 10 mg
76-965-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 76-961-0 Supplier Tocris Bioscience™ Supplier No. 7696/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective RET inhibitor

Selpercatinib, is a potent and selective receptor tyrosine kinase RET inhibitor (EC50 = 4 nM). It shows > 100-fold selectivity for > 95% of 228 purified kinase domains in radiometric assays. In vitro, it inhibits proliferation of cancer cell lines containing RET mutations or gene fusions. In vivo, it inhibits tumor growth in diverse RET mouse models. Selpercatinib is orally bioavailable.

Specifications

Chemical Name or Material 6-(2-Hydroxy-2-methylpropoxy)-4-[6-[6-[(6-methoxy-3-pyridinyl)methyl]-3,6-diazabicyclo[3.1.1 ]hept-3-yl]-3-pyridinyl]pyrazolo[1,5-a]pyridine-3-carbonitrile
CAS 2152628-33-4
Quantity 10 mg
Target Ret
Molecular Formula C29H31N7O3
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.