Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SELS Antibody (7F8G1) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP261876
Description
SELS Monoclonal specifically detects SELS in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SELS | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
ADO15, MGC104346, SBBI8, selenoprotein S, SelS, SEPS1, VCP-interacting membrane protein, VIMPMGC2553 | |
Mouse | |
21.2 kDa | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
7F8G1 | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
VIMP | |
Purified recombinant fragment of human VIMP (AA: 1-187) expressed in E. Coli. | |
Protein G purified | |
Signal Transduction | |
55829 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction