Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SELS Monoclonal Antibody (5G4A10)
GREENER_CHOICE

Catalog No. PIMA531951
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531951 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531951 Supplier Invitrogen™ Supplier No. MA531951
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31951 has been tested in indirect ELISA.

This gene encodes a transmembrane protein that is localized in the endoplasmic reticulum (ER). It is involved in the degradation process of misfolded proteins in the ER, and may also have a role in inflammation control. It participates in the transfer of misfolded proteins from the ER to the cytosol, where they are destroyed by the proteasome in a ubiquitin-dependent manner. This protein is a selenoprotein, containing the rare amino acid selenocysteine (Sec). Sec is encoded by the UGA codon, which normally signals translation termination. The 3' UTRs of selenoprotein mRNAs contain a conserved stem-loop structure, designated the Sec insertion sequence (SECIS) element, that is necessary for the recognition of UGA as a Sec codon, rather than as a stop signal.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SELS
Applications ELISA, Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 5G4A10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene SELENOS
Gene Accession No. Q9BQE4
Gene Alias 1500011E07Rik; AD-015; ADO15; C78786; chondroitin sulfate synthase 1; CHSY1; H4; H-4; H47; H-47; histocompatibility 47; MGC104346; MGC2553; minor histocompatibility antigen H47; SBBI8; selenoprotein S; Selenos; SELS; SEPS1; sg2; similar to selenoprotein S; Tanis; valosin-containing protein-interacting membrane protein; VCP interacting membrane selenoprotein; VCP-interacting membrane protein; VCP-interacting membrane selenoprotein; vimp; wu:fc16a03; zgc:136970
Gene Symbols SELENOS
Host Species Mouse
Immunogen Purified recombinant fragment of human VIMP (AA: 1-187) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55829
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.