Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SELS Mouse anti-Human, Polyclonal, Abnova™
Mouse polyclonal antibody raised against a full-length human SELS protein.
Supplier: Abnova Corporation H00055829B01P
Description
This gene encodes a selenoprotein, which contains a selenocysteine (Sec) residue at its active site. The selenocysteine is encoded by the UGA codon that normally signals translation termination. The 3′ UTR of selenoprotein genes have a common stem-loop structure, the sec insertion sequence (SECIS), that is necessary for the recognition of UGA as a Sec codon rather than as a stop signal. Studies suggest that this protein may regulate cytokine production, and thus play a key role in the control of the inflammatory response. Two alternatively spliced transcript variants encoding the same protein have been found for this gene. [provided by RefSeq]Specifications
SELS | |
Polyclonal | |
selenoprotein S | |
SELS | |
AD-015, ADO15, MGC104346, MGC2553, SBBI8, SEPS1, VIMP | |
Mouse | |
Affinity chromatography | |
RUO | |
55829 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC005840 | |
SELENOS | |
SELS (AAH05840, 1 a.a. ∽ 189 a.a) full-length human protein. | |
50 μg | |
Primary | |
Human | |
Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction