Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SELV Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563697
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563697 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563697 Supplier Invitrogen™ Supplier No. PA563697
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RTPTPLRTPT PVRTRTPIRA LTPVLTPSPA GTSPLVLTPA PAQIPTLVPT PALARIPRLV PPPAPAWIPT PVPTPVPVRN PT Highest antigen sequence identity to the following orthologs: Mouse - 47%, Rat - 52%.

This gene encodes a selenoprotein, which contains a selenocysteine (Sec) residue at its active site. The selenocysteine is encoded by the UGA codon that normally signals translation termination. The 3' UTR of selenoprotein genes have a common stem-loop structure, the sec insertion sequence (SECIS), that is necessary for the recognition of UGA as a Sec codon rather than as a stop signal.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SELV
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SELENOV
Gene Accession No. P59797
Gene Alias Selenoprotein V; SELENOV; SelV
Gene Symbols SELENOV
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SELV. Recombinant protein control fragment (Product #RP-99187).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 348303
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.