Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEMA3A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556149
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556149 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556149 Supplier Invitrogen™ Supplier No. PA556149
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VPRLKLSYKE MLESNNVITF NGLANSSSYH TFLLDEERSR LYVGAKDHIF SFDLVNIKDF QKIVWPVSYT RRDECKWAGK DILKECANFI KVL Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

This gene is a member of the semaphorin family and encodes a protein with an Ig-like C2-type domain, a PSI domain and a Sema domain. This secreted protein can function as either a chemorepulsive agent, inhibiting axonal outgrowth, or as a chemoattractive agent, stimulating the growth of apical dendrites. In both cases, the protein is vital for normal neuronal pattern development. Increased expression of this protein is associated with schizophrenia and is seen in a variety of human tumor cell lines. Also, aberrant release of this protein is associated with the progression of Alzheimer's disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEMA3A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SEMA3A
Gene Accession No. Q14563
Gene Alias COLL1; coll-1; collapsin 1; collapsin-1; HH16; Hsema-I; Hsema-III; MGC133243; sema D; sema domain, immunoglobulin domain (Ig), short basic domain, sec; sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3A; sema III; SEMA1; SEMA3A; Semad; SEMAIII; SEMAL; Semaphorin 3a; semaphorin D; semaphorin III; semaphorin L; semaphorin-3A; Semaphorin-D; SemD
Gene Symbols SEMA3A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SEMA3A. Recombinant protein control fragment (Product #RP-92278).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10371
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.