Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEMA3B Monoclonal Antibody (9C4F7)
GREENER_CHOICE

Catalog No. PIMA549294
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549294 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549294 Supplier Invitrogen™ Supplier No. MA549294
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human CACO-2 whole cell, human U87 whole cell, human K562 whole cell, rat kidney tissue, rat NRK whole cell, mouse kidney tissue, mouse ANA-1 whole cell. ICC/IF: Hela cell. Flow: HepG2 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

The semaphorin/collapsin family of molecules plays a critical role in the guidance of growth cones during neuronal development. The secreted protein encoded by this gene family member is important in axonal guidance and has been shown to act as a tumor suppressor by inducing apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEMA3B
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 9C4F7
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene SEMA3B
Gene Accession No. Q13214, Q62177
Gene Alias LUCA-1; Seam V antibody; SemA; sema A; sema A(V); sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3B; Sema V; SEMA3B; SEMA5; SEMAA; semaphorin 3B; semaphorin A; semaphorin-3B; semaphorin-A; semaphorin-V; semaV
Gene Symbols SEMA3B
Host Species Mouse
Immunogen E.coli-derived human Semaphorin 3B/SEMA3B recombinant protein (Position: Y55-R624).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20347, 363142, 7869
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.