Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEMA3C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547189
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547189 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547189 Supplier Invitrogen™ Supplier No. PA547189
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

In Western blots, less than 5% cross-reactivity with recombinant human (rh) Semaphorin 3A, rhSemaphorin 3B, rhSemaphorin 6A, recombinant mouse (rm) Semaphorin 3E, rmSemaphorin 3F, rmSemaphorin 6A, and rmSemaphorin 7A is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

The SEMA3C gene encodes the protein semaphorin 3C, a member of the semaphorin family known for its roles in axonal guidance, cellular signaling, and vascular development. Semaphorin 3C is involved in guiding neurons during development by serving as a repulsive cue, helping establish proper neural network formation through interaction with neuropilin and plexin receptors. It also plays crucial roles in angiogenesis and cardiac development by influencing endothelial cell behavior and migration. In the context of cancer, SEMA3C is implicated in tumor progression, where its dysregulation can promote tumor cell invasion and metastasis by modifying the extracellular matrix and affecting cellular adhesion. Research has demonstrated that SEMA3C can alter the tumor microenvironment, making it a potential target for cancer therapies aimed at inhibiting metastasis and controlling tumor growth. Understanding SEMA3C's function in various biological processes continues, highlighting its significance in developmental biology and disease modulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEMA3C
Applications Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene SEMA3C
Gene Accession No. Q62181
Gene Alias 1110036B02Rik; sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3C; sema E; Sema3c; Semae; semaphorin 3C; semaphorin E; semaphorin-3C; Semaphorin-E; SemE
Gene Symbols SEMA3C
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Semaphorin 3C Gln24-Ser751 (Arg48Ala, Arg52Ala).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20348
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.