Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sema3C Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987433
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7433 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7433 Supplier Bioss Supplier No. BS6692R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SEMA3C may be involved in diverse cell survival mechanisms.

Specifications

Antigen Sema3C
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SEMA3C
Gene Accession No. Q99985
Gene Alias 1110036B02Rik; sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3C; Sema E; Sema3c; SEMAE; semaphorin 3C; semaphorin E; Semaphorin-3C; Semaphorin-E; SemE
Gene Symbols SEMA3C
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Sema3C.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10512
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.