Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEMA3D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557986
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557986 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557986 Supplier Invitrogen™ Supplier No. PA557986
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LYSGTASDFL GKDTAFTRSL GPTHDHHYIR TDISEHYWLN GAKFIGTFFI PDTYNPDDDK IYFFFRESSQ EGSTSDKTIL SRVGRVCKND VG Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 93%.

SEMA3D induces the collapse and paralysis of neuronal growth cones. SEMA3D could potentially act as repulsive cues toward specific neuronal populations.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEMA3D
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SEMA3D
Gene Accession No. O95025
Gene Alias 4631426B19Rik; coll-2; collapsin 2; sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3D; SEMA3D; semaphorin 3D; semaphorin, homolog of the avian Sema3D; semaphorin-3D; Sema-Z2; UNQ760/PRO1491
Gene Symbols SEMA3D
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SEMA3D. Recombinant protein control fragment (Product #RP-94995).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 223117
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.