Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEMA3E/Semaphorin 3E Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981402
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1402 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1402 Supplier Bioss Supplier No. BS11480R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The Semaphorins make up the largest family of axon guidance cues yet described. Semaphorins are divided into 8 classes (classes 3-7 found in vertebrates). Class 3 Semaphorins are secreted, classes 4 through 6 are transmembrane proteins, and class 7 are membrane associated via glycosylphosphatidylinositol (GPI) linkage. They are characterized structurally by a conserved approximately 400 amino acid sema domain. They are classically described as collapsing factors and mediators of axon repulsion, although they may also act as context-dependent chemoattractants. Semaphorins have been shown to have roles in cardiovascular development and in the regulation of immune cell antigen presentation. Receptors or receptor complexes that mediate semaphorin signaling include proteins of the Neuropilin and Plexin families.

Specifications

Antigen SEMA3E/Semaphorin 3E
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SEMA3E
Gene Accession No. O15041
Gene Alias 6430702L12; AA408817; coll-5; KIAA0331; mKIAA0331; M-sema H; M-SEMAH; M-SemaK; sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3E; Sema H; Sema3e; SEMAH; semaphorin 3E; Semaphorin-3E; Semaphorin-H; Semh
Gene Symbols SEMA3E
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SEMA3E/Semaphorin 3E.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9723
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.