Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEMA3G Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550261
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550261 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550261 Supplier Invitrogen™ Supplier No. PA550261
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total SEMA3G protein.

Has chemorepulsive activities for sympathetic axons. Ligand of NRP2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEMA3G
Applications Western Blot
Classification Polyclonal
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene SEMA3G
Gene Accession No. Q9NS98
Gene Alias sem2; SEMA3G; semaphorin 3G; Semaphorin sem2; Semaphorin-3G
Gene Symbols SEMA3G
Host Species Rabbit
Immunogen Fusion protein corresponding to residues near the C terminal of human sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3G.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56920
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.