Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SEMA4A Polyclonal Antibody
Rabbit Polyclonal Antibody
$589.50
Specifications
Antigen | SEMA4A |
---|---|
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5111376
|
Invitrogen™
PA5111376 |
100 μL |
Each for $589.50
|
|
|||||
PIPA5111608
|
Invitrogen™
PA5111608 |
100 μL |
Each for $589.50
|
|
|||||
Description
Immunogen sequence: LPFNVIRHAV LLPADSPTAP HIYAVFTSQW QVGGTRSSAV CAFSLLDIER VFKGKYKELN KETSRWTTYR GPETNPRPGS CSVGPSSD.
SEMA4A is a member of the semaphorin family of soluble and transmembrane proteins. Semaphorins are involved in guidance of axonal migration during neuronal development and in immune responses.Specifications
SEMA4A | |
Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H3S1 | |
64218 | |
Recombinant Protein corresonding to Human SEMA4A. Recombinant protein control fragment (Product #RP-104975). | |
Antibody |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
SEMA4A | |
AI132332; CORD10; RP35; Sema B; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4A; Sema4a; Semab; semaphorin 4A; semaphorin B; semaphorin-4A; Semaphorin-B; SemB; UNQ783/PRO1317 | |
SEMA4A | |
Primary | |
Antigen Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title