Learn More
Invitrogen™ SEMA4A Polyclonal Antibody

Description
Immunogen sequence: PNLNSWKQDM ERGNPEWACA SGPMSRSLRP QSRPQIIKEV LAVPNSILEL PCPHLSALAS YYWSHGPAAV PEASSTVYNG SLLLIVQDGV GGLYQCWATE NGFSYPVISY WVDSQDQTLA LDPELAGIPR EHVKVPLTRV SGGA Highest antigen sequence indentity to the following orthologs: Rat - 81%, Mouse - 79%.
Specifications
Specifications
| Antigen | SEMA4A |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | SEMA4A |
| Gene Accession No. | Q9H3S1 |
| Gene Alias | AI132332; CORD10; RP35; Sema B; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4A; Sema4a; Semab; semaphorin 4A; semaphorin B; semaphorin-4A; Semaphorin-B; SemB; UNQ783/PRO1317 |
| Gene Symbols | SEMA4A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.