Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SEMA4B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA533017
Description
Percent identity with other species by BLAST analysis: Human (100%) Gorilla, Gibbon, Monkey (94%) Marmoset, Hamster (88%) Elephant, Rabbit (82%).
Inhibits axonal extension by providing local signals to specify territories inaccessible for growing axons. [UniProt].
Specifications
| SEMA4B | |
| Polyclonal | |
| Unconjugated | |
| SEMA4B | |
| KIAA1745; mKIAA1745; sema C; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4B; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, 4B; Sema4b; SEMAC; semaphorin 4B; semaphorin-4B; Semaphorin-C; SemC; UNQ749/PRO1480 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10509 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.25 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q9NPR2 | |
| SEMA4B | |
| Synthetic 17 amino acid peptide from internal region of human SEMA4B. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction