Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SEMA4B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110780
Description
Immunogen sequence: EAEHISNYTA LLLSRDGRTL YVGAREALFA LSSNLSFLPG GEYQELLWGA DAEKKQQCSF KGKDPQRDCQ NYIKILLPLS GSHLFTCGTA AFSPMCTYIN MENFTLARDE KGNVLLEDGK GRCPFDPNFK STALVVDGEL YTGTVSSF.
Inhibits axonal extension by providing local signals to specify territories inaccessible for growing axons. [UniProt].
Specifications
SEMA4B | |
Polyclonal | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NPR2 | |
SEMA4B | |
Recombinant Protein corresonding to Human SEMA4B. Recombinant protein control fragment (Product #RP-90547). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
SEMA4B | |
KIAA1745; mKIAA1745; sema C; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4B; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, 4B; Sema4b; SEMAC; semaphorin 4B; semaphorin-4B; Semaphorin-C; SemC; UNQ749/PRO1480 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10509 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction