Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SEMA4C Polyclonal Antibody
Sheep Polyclonal Antibody
Supplier: Invitrogen™ PA547812
Description
In direct ELISAs, approximately 20% cross-reactivity with recombinant human Semaphorin 4C and less than 5% cross-reactivity with recombinant mouse (rm) Semaphorin 4B, rmSemaphorin 4F, and Semaphorin 4G is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
SEMA4C (semaphorin-4C), also known as SEMAF, is an 833 amino acid single-pass type I membrane protein that contains one sema domain, one PSI domain and one Ig-like C2-type domain. Expressed in a variety of tissues, including lung, kidney and brain, SEMA4C interacts with GIPC and NCDN and is thought to play a role in the formation of neural networks during development.
Specifications
SEMA4C | |
Polyclonal | |
Unconjugated | |
SEMA4C | |
AI426163; FLJ20369; KIAA1739; MGC126382; MGC126383; M-Sema F; M-SEMA-F; RGD1562837; S4c; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4C; Sema I; SEMA4C; SEMACL1; SEMAF; SEMAI; semaphorin 4C; semaphorin-4C; Semaphorin-C-like 1; UNQ5855/PRO34487 | |
Sheep | |
Antigen affinity chromatography | |
RUO | |
20353, 301346 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q64151 | |
SEMA4C | |
Mouse myeloma cell line NS0-derived recombinant mouse Semaphorin 4C Glu22-Gly664. | |
100 μg | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction