Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEMA4C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA576357
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA576357 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA576357 Supplier Invitrogen™ Supplier No. PA576357
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

SEMA4C (semaphorin-4C), also known as SEMAF, is an 833 amino acid single-pass type I membrane protein that contains one sema domain, one PSI domain and one Ig-like C2-type domain. Expressed in a variety of tissues, including lung, kidney and brain, SEMA4C interacts with GIPC and NCDN and is thought to play a role in the formation of neural networks during development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEMA4C
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene SEMA4C
Gene Accession No. Q64151, Q9C0C4
Gene Alias AI426163; FLJ20369; KIAA1739; MGC126382; MGC126383; M-Sema F; M-SEMA-F; RGD1562837; S4c; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4C; Sema I; SEMA4C; SEMACL1; SEMAF; SEMAI; semaphorin 4C; semaphorin-4C; Semaphorin-C-like 1; UNQ5855/PRO34487
Gene Symbols SEMA4C
Host Species Rabbit
Immunogen Recombinant fusion protein of human SEMA4C.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20353, 54910
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.