Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SEMA4D Monoclonal Antibody (OTI6G1), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA809848

Description
SEMA4D (CD100) is a cell surface receptor for PLXN1B and PLXNB2. It plays an important role in cell-cell signaling and promotes reorganization of the actin cytoskeleton. CD100 also plays a role in axonal growth cone guidance in the developing central nervous system, as well as, regulates dendrite and axon branching and morphogenesis. CD100 is also active in the immune system - it induces B-cells to aggregate and improves thru viability.Specifications
SEMA4D | |
Monoclonal | |
Unconjugated | |
SEMA4D | |
C9orf164, CD100, COLL4, M-sema-G, SEMAJ, coll-4 | |
Mouse | |
Affinity Chromatography | |
RUO | |
10507 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
OTI6G1 | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q92854 | |
SEMA4D | |
Full length human recombit protein of human SEMA4D produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction