Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEMA4D Mouse anti-Human, Unconjugated, Clone: A8, Abnova™

Catalog No. 89337634 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-634 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-634 Supplier Abnova Corporation Supplier No. MAB21141
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

SEMA4D Monoclonal antibody specifically detects SEMA4D in Human samples. It is validated for IF, Flow Cyt.

Specifications

Antigen SEMA4D
Applications Immunofluorescence, Flow Cytometry
Classification Monoclonal
Clone A8
Conjugate Unconjugated
Description sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4D
Gene SEMA4D
Gene Alias C9orf164, CD100, FLJ33485, FLJ34282, FLJ39737, FLJ46484, M-sema-G, MGC169138, MGC169141, SEMAJ, coll-4
Host Species Mouse
Immunogen PHA stimulated human peripheral blood lymphocytes
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 10507
Target Species Human
Content And Storage Store at 4°C
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.