Learn More
SEMA4D Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304395AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse
Semaphorins are neuronal chemorepellants that direct pioneering neurons during nervous system development. CD100 is a novel semaphorin that induces B cells to aggregate and improves their viability in vitro. CD100 modifies CD40-CD40L B-cell signaling by augmenting B-cell aggregation and survival and down-regulating CD23 expression. CD72 is a lymphocyte receptor for the Class IV semaphorin CD100, which is involved in regulating B cell signaling.Specifications
SEMA4D | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q92854 | |
SEMA4D | |
Between 812 and 862 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
SEMA4D | |
Semaphorin-4D, Leukocyte activation antigen CD100, BB18, A8, GR3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10507 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.