Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEMA4F Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542878
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542878 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542878 Supplier Invitrogen™ Supplier No. PA542878
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: PFSGERPRRI DWMVPEAHRQ NCRKKGKKEG DLGGRKTLQQ RWTTFLKADL Sequence homology: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

SEMA4F has growth cone collapse activity against retinal ganglion-cell axons.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEMA4F
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SEMA4F
Gene Accession No. O95754
Gene Alias M-SEMA; m-Sema-M; PRO2353; S4F; sema domain immunoglobulin domain transmembrane domain (TM) and short cytoplasmic domain (semaphorin) 4F; sema domain, immunoglobulin domain (Ig), TM domain, and short cy; sema domain, immunoglobulin domain (Ig), TM domain, and short cytoplasmic domain; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4F; Sema M; Sema W; Sema4f; SEMAM; semaphorin 4f; semaphorin M; semaphorin W; semaphorin-4F; Semaphorin-M; semaphorin-W; SEMAW; ssemaphorin 4F
Gene Symbols SEMA4F
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human SEMA4F.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10505
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.