Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEMA4F Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566814
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566814 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566814 Supplier Invitrogen™ Supplier No. PA566814
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RTSLPISEADS CLTRFAVPHT YNYSVLLVDP ASHTLYVGAR DTIFALSLPF SGERPRRIDW MVPEAHRQNC RKKGKKEDEC HNFVQIL Highest antigen sequence identity to the following orthologs - mouse 91%, rat 91%.

SEMA4F has growth cone collapse activity against retinal ganglion-cell axons.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEMA4F
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SEMA4F
Gene Accession No. O95754
Gene Alias M-SEMA; m-Sema-M; PRO2353; S4F; sema domain immunoglobulin domain transmembrane domain (TM) and short cytoplasmic domain (semaphorin) 4F; sema domain, immunoglobulin domain (Ig), TM domain, and short cy; sema domain, immunoglobulin domain (Ig), TM domain, and short cytoplasmic domain; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4F; Sema M; Sema W; Sema4f; SEMAM; semaphorin 4f; semaphorin M; semaphorin W; semaphorin-4F; Semaphorin-M; semaphorin-W; SEMAW; ssemaphorin 4F
Gene Symbols SEMA4F
Host Species Rabbit
Immunogen Recombinant Human SEMA4F. Recombinant protein control fragment (Product #RP-105375).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10505
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.