Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SEMA4F Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584626
Description
Immunogen sequence: DVAVLRPELG AGTPIFYGIF SSQWEGATIS AVCAFRPQDI RTVLNGPFRE LKHDCNRGLP VVDNDVPQPR.
SEMA4F has growth cone collapse activity against retinal ganglion-cell axons.
Specifications
SEMA4F | |
Polyclonal | |
Unconjugated | |
SEMA4F | |
M-SEMA; m-Sema-M; PRO2353; S4F; sema domain immunoglobulin domain transmembrane domain (TM) and short cytoplasmic domain (semaphorin) 4F; sema domain, immunoglobulin domain (Ig), TM domain, and short cy; sema domain, immunoglobulin domain (Ig), TM domain, and short cytoplasmic domain; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4F; Sema M; Sema W; Sema4f; SEMAM; semaphorin 4f; semaphorin M; semaphorin W; semaphorin-4F; Semaphorin-M; semaphorin-W; SEMAW; ssemaphorin 4F | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10505 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O95754 | |
SEMA4F | |
Recombinant protein corresponding to Human SEMA4F. Recombinant protein control fragment (Product #RP-108496). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction