Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEMA4F Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981399
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1399 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1399 Supplier Bioss Supplier No. BS11478R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SEMA4F has growth cone collapse activity against retinal ganglion-cell axons.

Specifications

Antigen SEMA4F
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SEMA4F
Gene Accession No. O95754
Gene Alias BOS_11353; M-SEMA; m-Sema-M; PRO2353; S4F; sema domain immunoglobulin domain transmembrane domain (TM) and short cytoplasmic domain (semaphorin) 4F; sema domain, immunoglobulin domain (Ig), TM domain, and short cy; sema domain, immunoglobulin domain (Ig), TM domain, and short cytoplasmic domain; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4F; Sema M; Sema W; Sema4f; SEMAM; semaphorin 4f; semaphorin M; semaphorin W; Semaphorin-4F; Semaphorin-M; Semaphorin-W; Semaw; ssemaphorin 4F
Gene Symbols SEMA4F
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SEMA4F.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10505
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.