Learn More
Invitrogen™ SEMA4G Polyclonal Antibody
Sheep Polyclonal Antibody
Supplier: Invitrogen™ PA547847
Description
In direct ELISAs, approximately 20% cross-reactivity with recombinant human Semaphorin 4G is observed, and less than 1% cross-reactivity with recombinant mouse (rm) Semaphorin 4B, rmSemaphorin 4C, and rmSemaphorin 4F is observed. Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.
The SEMA4G gene encodes semaphorin 4G, a member of the semaphorin protein family known for their roles in cellular guidance, signaling, and development. Semaphorin 4G is involved in axonal guidance, where it influences the direction and growth of neurons by interacting with receptor complexes such as plexins. This interaction is critical in neural development, contributing to the proper formation of neural circuits by ensuring that neurons connect accurately and functionally. Beyond its role in the nervous system, SEMA4G also participates in immune responses, where it modulates the activity and migration of immune cells, influencing processes like inflammation and immune surveillance. This protein's modulation of cell-to-cell communication and migration extends to its implications in cancer, where altered semaphorin signaling can impact tumor progression and metastasis through changes in cell adhesion and migration properties. Research into SEMA4G continues to investigate its precise mechanisms and interactions, providing insights into therapeutic opportunities in neural disorders and cancer.
Specifications
SEMA4G | |
Polyclonal | |
Unconjugated | |
SEMA4G | |
AI507908; AW554132; KIAA1619; KIAA1619FLJ20590; MGC102867; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4G; Sema4g; semaphorin 4G; semaphorin-4G | |
Sheep | |
Antigen affinity chromatography | |
RUO | |
26456 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Frozen), Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q9WUH7 | |
SEMA4G | |
Mouse myeloma cell line NS0-derived recombinant mouse Semaphorin 4G Val18-Met673. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.