Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEMA5A Monoclonal Antibody (C3), Invitrogen™

Catalog No. FEMA551132
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA551132 20 μL
FEMA551131 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEMA551132 Supplier Invitrogen™ Supplier No. MA551132
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Members of the semaphorin protein family, such as SEMA5A, are involved in axonal guidance during neural development (Adams et al., 1996 [PubMed 8817451]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEMA5A
Applications Western Blot, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone C3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene SEMA5A
Gene Accession No. D3ZTD8, Q13591
Gene Alias 5930434A13; 9130201M22Rik; AI464145; Ensembl:ENSRNOP00000016506}; M-Sema D; sema domain, seven thrombospondin repeats (type 1 and type 1-like), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 5A; sema F; SEMA5A; sema5a {ECO:0000312; SEMAF; semaphorin 5A; semaphorin F; semaphorin-5A; semaphorin-5A {ECO:0000250; Semaphorin-F; semF; UniProtKB:Q13591}
Gene Symbols SEMA5A
Host Species Mouse
Immunogen Recombinant Human SEMA5A protein, Val319-Gln668 (Accession #Q13591).
Purification Method Protein A/G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 310207, 9037
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.