Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEMA5A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531283
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531283 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531283 Supplier Invitrogen™ Supplier No. PA531283
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Mouse brain. Predicted reactivity: Mouse (95%), Rat (94%), Rhesus Monkey (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Members of the semaphorin protein family, such as SEMA5A, are involved in axonal guidance during neural development (Adams et al., 1996 [PubMed 8817451]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEMA5A
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene SEMA5A
Gene Accession No. Q13591, Q62217
Gene Alias 5930434A13; 9130201M22Rik; AI464145; Ensembl:ENSRNOP00000016506}; M-Sema D; sema domain, seven thrombospondin repeats (type 1 and type 1-like), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 5A; sema F; SEMA5A; sema5a {ECO:0000312; SEMAF; semaphorin 5A; semaphorin F; semaphorin-5A; semaphorin-5A {ECO:0000250; Semaphorin-F; semF; UniProtKB:Q13591}
Gene Symbols SEMA5A
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the Extracellular domain of human Semaphorin 5A. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20356, 9037
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.