Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEMA5A Polyclonal Antibody, Invitrogen™

Catalog No. PIPA547791
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547791 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547791 Supplier Thermo Scientific Supplier No. PA547791

Sheep Polyclonal Antibody

In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) Sema3B, rhSema4C, and rhSema6A is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

SEMA5A Polyclonal antibody specifically detects SEMA5A in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEMA5A
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene SEMA5A
Gene Accession No. D3ZTD8, Q13591, Q62217
Gene Alias 5930434A13; 9130201M22Rik; AI464145; Ensembl:ENSRNOP00000016506}; M-Sema D; sema domain, seven thrombospondin repeats (type 1 and type 1-like), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 5A; sema F; SEMA5A; sema5a {ECO:0000312; SEMAF; semaphorin 5A; semaphorin F; semaphorin-5A; semaphorin-5A {ECO:0000250; Semaphorin-F; semF; UniProtKB:Q13591}
Gene Symbols SEMA5A
Host Species Sheep
Immunogen CHO-derived recombinant human Semaphorin 5A Glu23-Thr765
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20356, 310207, 9037
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.