Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SEMA5B Polyclonal Antibody
Rabbit Polyclonal Antibody
$237.00 - $539.50
Specifications
Antigen | SEMA5B |
---|---|
Concentration | 0.81 mg/mL |
Content And Storage | -20°C |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119406
|
Invitrogen™
PA5119406 |
200 μL |
Each for $539.50
|
|
|||||
PIPA5121622
|
Invitrogen™
PA5121622 |
20 μL |
Each for $237.00
|
|
|||||
Description
SEMA5B Polyclonal Antibody for Western Blot, IHC (P)
May act as positive axonal guidance cues.Specifications
SEMA5B | |
-20°C | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
SEMA5B | |
AI893641; Kiaa1445; mKIAA1445; sema domain, seven thrombospondin repeats (type 1 and type 1-like), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 5B; sema G; Sema5b; Semag; semaphorin 5B; semaphorin-5B; Semaphorin-G; SemG; UNQ5867/PRO34001 | |
SEMA5B | |
Primary | |
Antigen Affinity Chromatography |
0.81 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Q9P283 | |
54437 | |
Recombinant protein Semaphorin 5B. The antigen corresponds to amino acid range 36-161 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title