Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEMA6A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143762
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143762 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143762 Supplier Invitrogen™ Supplier No. PA5143762
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects a 120kDa* protein corresponding to the apparent molecular mass of Sema-6A on SDS-PAGE immunoblots of Sema-6A transfected COS7 cells, human colorectal cancer cells, and rat PC12 cells. In addition, shRNA knockdown of Sema-6A expression significantly reduces detection of Sema-6A using anti-Sema-6A (C-terminus) antibody.

SEMA6A can act as repulsive axon guidance cues. It may play a role in channeling sympathetic axons into the sympathetic chains and controlling the temporal sequence of sympathetic target innervation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEMA6A
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gene SEMA6A
Gene Accession No. O35464, Q9H2E6
Gene Alias 9330158E07; A730020P05Rik; AI851735; b2b997Clo; HT018; KIAA1368; Mutant line 997; SEMA; sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6A; sema Q; Sema VIA; SEMA6A; SEMA6A1; SEMA6A-1; semaphorin 6A; semaphorin 6A-1; semaphorin Q; Semaphorin VIA; semaphorin-6A; Semaphorin-6A-1; Semaq; VIA
Gene Symbols SEMA6A
Host Species Rabbit
Immunogen Semaphorin 6A synthetic peptide (coupled to carrier protein) corresponding to amino acids in the C-terminus of human Sema-6A. The sequence used is identical to similar regions in rat and mouse Sema-6A, and has low homology to other semaphorin family members.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20358, 361324, 57556
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.