Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEMA6B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563176
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563176 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563176 Supplier Invitrogen™ Supplier No. PA563176
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AGTVLKFLVR PNASTSGTSG LSVFLEEFET YRPDRCGRPG GGETGQRLLS LELDAASGGL LAAFPRCVVR VPVARCQQYS GCMKNCIG Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 87%.

The SEMA6B gene encodes semaphorin 6B, which belongs to the semaphorin family of proteins known for their roles in guiding axonal development and modulating cellular signaling pathways. Semaphorin 6B primarily functions in the nervous system where it influences axonal guidance, neuronal development, and synapse formation through interactions with neuropilin and plexin receptor complexes. These interactions are crucial during neural development, ensuring proper neuronal wiring and the establishment of functional neural circuits. In addition to its key role in neural connectivity, SEMA6B is increasingly recognized for its involvement in immune responses, where it can modulate the activity and migration of immune cells. In cancer biology, altered expression of SEMA6B has been implicated in tumor progression, affecting tumor cell adhesion and migration, thereby influencing metastasis. Research continues to explore the specific mechanisms of SEMA6B's action and how it can be targeted for therapeutic interventions in neural and cancer-related diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEMA6B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SEMA6B
Gene Accession No. Q9H3T3
Gene Alias Sema; sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6B; sema N; Sema VIB; sema Z; SEMA6B; SEMAN; semaphorin 6B; semaphorin VIB; semaphorin Z; semaphorin-6B; semaphorin-6Ba; semaphorin-N; Semaphorin-Z; SEMA-VIB; semaZ; SEM-SEMA-Y; UNQ1907/PRO4353; VIb
Gene Symbols SEMA6B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SEMA6B. Recombinant protein control fragment (Product #RP-103230).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10501
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.