Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEMA6D Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985033
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5033 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5033 Supplier Bioss Supplier No. BS2943R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Semaphorins are a large family, including both secreted and membrane associated proteins, many of which have been implicated as inhibitors or chemorepellents in axon pathfinding, fasciculation and branching, and target selection. All semaphorins possess a semaphorin (Sema) domain and a PSI domain (found in plexins, semaphorins and integrins) in the N-terminal extracellular portion. Additional sequence motifs C-terminal to the semaphoring domain allow classification into distinct subfamilies. Results demonstrate that transmembrane semaphorins, like the secreted ones, can act as repulsive axon guidance cues. SEMA6D is a class 6 vertebrate transmembrane semaphorin that demonstrates alternative splicing. Six transcript variants have been identified and expression of the distinct encoded isoforms is thought to be regulated in a tissue- and development-dependent manner.

Specifications

Antigen SEMA6D
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SEMA6D
Gene Accession No. Q8NFY4
Gene Alias 1110067B02Rik; AA409156; BOS_10967; D330011G23; FLJ11598; hypothetical protein LOC406272; Kiaa1479; KIAA1479FLJ11598; mKIAA1479; sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6D; Sema6d; sema6d protein; semaphorin 6D; semaphorin-6C; Semaphorin-6D; wu:fi06a12; wu:fi76a10; zgc:56170
Gene Symbols SEMA6D
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SEMA6D.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80031
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.